Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
Empirical Formula (Hill Notation):
C6H13NO7
CAS Number:
Molecular Weight:
211.17
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12352201
EC Number:
238-051-8
MDL number:
Pricing and availability is not currently available.
form
solid
Quality Segment
SMILES string
OC[C@@H](O)[C@@H](O)[C@H](O)[C@H](O)C[N+]([O-])=O
InChI
1S/C6H13NO7/c8-2-4(10)6(12)5(11)3(9)1-7(13)14/h3-6,8-12H,1-2H2
InChI key
HOFCJTOUEGMYBT-UHFFFAOYSA-N
Application
1-Deoxy-1-nitro-D-mannitol (DN-Man) is use as a voltammetric, in differential pulse voltammetry (DPV), reagent for monitoring blood-brain barrier (BBB) transport.
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.