Skip to Content
Merck

Skip To

D200

N,N-Dimethyl-1-phenyl-2-ethanamine hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C10H15N · HCl
Molecular Weight:
185.69
UNSPSC Code:
12352100
PubChem Substance ID:
EC Number:
219-511-7
MDL number:
Form:
solid
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

solid

drug control

USDEA Schedule I; Home Office Schedule 1; regulated under CDSA - not available from Sigma-Aldrich Canada

solubility

H2O: soluble

SMILES string

Cl.CN(C)CCc1ccccc1

InChI

1S/C10H15N.ClH/c1-11(2)9-8-10-6-4-3-5-7-10;/h3-7H,8-9H2,1-2H3;1H

InChI key

OVBRYLBVLSAOAS-UHFFFAOYSA-N



pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content


Filiberto Mastrangelo et al.
The International journal of oral & maxillofacial implants, 35(2), 320-329 (2020-03-07)
The aim of this study was to compare how two innovative laser titanium surfaces and sandblasted and acid-etched surfaces influence human osteoblast behavior during osteogenesis and the initial phases of bone deposition. Human osteoblasts from human adipose stem cells were



Global Trade Item Number

SKUGTIN
GILF171300-10X96EA04065274079516
GILF161930-10000EA04065274079110
GILF167103-960EA04065274079264
GILF161931-5X200EA04065274079127
GILF171302-2EA04065274079530
GILF167013-960EA04065274079219
GILF172300-10X96EA04065274079752
GILF171204-2EA04065274079509

Questions

Reviews

No rating value

Active Filters