Skip to Content
Merck

Skip To

D1444

6,2′-Dihydroxyflavone

≥98% (HPLC), powder

Synonym(s):

6-hydroxy-2-(2-hydroxyphenyl)-4H-1-benzopyran-4-one, DHF

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C15H10O4
Molecular Weight:
254.24
NACRES:
NA.77
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder


assay

≥98% (HPLC)

form

powder

color

white

solubility

DMSO: >10 mg/mL

storage temp.

room temp

Biochem/physiol Actions

6,2′-Dihydroxyflavone is a partial inverse agonist at GABAA receptors with α-subunit selectivity.

Features and Benefits

This compound is featured on the GABAA Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.


pictograms

Skull and crossbonesCorrosion

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Articles

We offer many products related to GABAA receptors for your research needs.


Feng Wang et al.
Neuropharmacology, 53(4), 574-582 (2007-08-08)
Neuroactivity of a number of flavonoids is mediated by modulation of type A gamma-aminobutyric acid (GABA(A)) receptor function via benzodiazepine sites, mostly as partial agonists. In the present study, 6,2'-dihydroxyflavone (DHF) was characterized for potential inverse agonistic activity, and its



Global Trade Item Number

SKUGTIN
D1444-25MG04061832260211
D1444-5MG04061832071886

Questions

Reviews

No rating value

Active Filters