Skip to Content
Merck

Skip To

D0761

Diacetoxyscirpenol

from Fusarium sambucinum

Synonym(s):

12,13-Epoxytrichothec-9-ene-3,4,15-triol-4,15-diacetate, 4β,15-Diacetoxy-3α-hydroxy-12,13 epoxy-trichothec-9-ene, Anguidin

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C19H26O7
CAS Number:
Molecular Weight:
366.41
PubChem Substance ID:
UNSPSC Code:
12352200
EC Number:
218-873-3
MDL number:
Beilstein/REAXYS Number:
1354986


biological source

Fusarium sambucinum

storage temp.

2-8°C

SMILES string

CC(=O)OC[C@]12CCC(C)=C[C@H]1O[C@@H]3[C@H](O)[C@@H](OC(C)=O)[C@@]2(C)[C@@]34CO4



pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 2 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



A Johannisson et al.
Cell biology and toxicology, 15(4), 203-215 (2000-03-04)
Four trichothecene mycotoxins--the type A trichothecenes T2-toxin and diacetoxyscirpenol and the type B trichothecenes nivalenol and deoxynivalenol--were studied. The effects of these mycotoxins on the expression of the sequentially expressed activation markers CD69, CD25, and CD71 and on proliferation of
M A Qureshi et al.
Immunopharmacology and immunotoxicology, 20(4), 541-553 (1998-11-07)
4 beta, 15 Diacetoxyscirpenol (DAS) mycotoxin produced by Fusarium species was tested for detrimental effects on macrophage viability, phagocytosis, and Fc-receptor expression. Sephadex-elicited chicken abdominal cells were harvested to establish adherent macrophage monolayers on glass coverslips. Coverslips were then assigned
J Brake et al.
Poultry science, 78(12), 1690-1694 (2000-01-08)
Two experiments were conducted to determine the effects of 4,15-diacetoxyscirpenol (DAS) on fertility and hatchability of broiler breeders. In Experiment 1, naturally mated broiler breeders were studied. A limited daily allocation of feed containing 0 (basal), 1.25, 2.5, or 5.0