CMC0009
STEADY Electrocompetent Cells
Escherichia coli, rod shaped
About This Item
Recommended Products
Product Name
STEADY Electrocompetent Cells, for plasmid production using unstable DNA
biological source
Escherichia coli
grade
Molecular Biology
form
buffered aqueous solution
growth mode
adherent or suspension
morphology
rod shaped
technique(s)
microbiological culture: suitable
cell transformation
competent cell type: electrocompetent
transformation efficiency: ≥1 x 1010 cfu/μg
shipped in
dry ice
storage temp.
−70°C
General description
Clones with inverted repeats or other sequnces prone to recombination are commonly found in retroviral genes, and require cells such as this product to be propagated stably.
Genotype
recA13 supE44 ara-14 galK2 lacY1 proA2 rpsL20(StrR) xyl-5 λ– leu mtl-1 F– mcrB mrr hsdS20(rB–, mB–)
Features and Benefits
- Clone unstable sequences with reliable results
- Available as chemically competent or electrocompetent cells
- Your choice of two packaging formats
- High efficiency: over 1 × 108 cfu/μg (chem) or 1 × 1010 cfu/μg (electro)
- Excellent value
Components
- STEADY electrocompetent cells
- pUC 19 transformation control DNA
- recovery medium for cloning
related product
Storage Class Code
10 - Combustible liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service