Skip to Content
Merck

Skip To

CMC0004

SIG10 MAX Electrocompetent Cells

Escherichia coli, rod shaped

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
41106202
NACRES:
NA.85
Biological source:
Escherichia coli
Growth mode:
adherent or suspension
Morphology:
rod shaped


Product Name

SIG10 MAX Electrocompetent Cells, for general cloning & library production

biological source

Escherichia coli

grade

Molecular Biology

form

buffered aqueous suspension

growth mode

adherent or suspension

morphology

rod shaped

technique(s)

microbiological culture: suitable

cell transformation

competent cell type: electrocompetent
transformation efficiency: ≥2 x 1010 cfu/μg

shipped in

dry ice

storage temp.

−70°C

General description

The SIG10 MAX Electrocompetent Cells provide large numbers of transformants from hard-to-clone fragments or limited DNA at a lower price.

Genotype

F- mcrA Δ(mrr-hsdRMS-mcrBC) endA1 recA1 Φ80dlacZΔM15 ΔlacX74 araD139 Δ(ara,leu)7697galU galK rpsL nupG λ- tonA (StrR)

Features and Benefits

  • share the most useful genetic elements of standard cloning strains like DH5a, DH10B, JM109, TOP10, etc. and directly replace them in cloning protocols.
  • incorporate a unique manufacturing technology that increases transformation efficiency, recombinant yields and reliability.
  • provide solutions for a wide range of applications at economical prices.
  • Blue - white screening

Other Notes

  • SIG10 MAX electrocompetent cells
  • pUC 19 transformation control DNA
  • recovery medium for cloning



Storage Class

10 - Combustible liquids

WGK

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters