Skip to Content
Merck
All Photos(1)

Key Documents

05-1530-M

Sigma-Aldrich

Anti-Importin beta-1 Antibody, clone 3H14

clone 3H14, from mouse

Synonym(s):

Karyopherin subunit beta-1, Nuclear factor P97, importin 1, importin 90, importin beta-1 subunit, karyopherin (importin) beta 1, karyopherin beta 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€421.00

€421.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
€421.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€421.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3H14, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... KPNB1(3837)

General description

Importin beta-1 (also known as Importin subunit beta-1) facilitates nuclear protein import. This occurs either though association with a nuclear localization signal (NLS) and an adapter protein , such as an importin-alpha subunit, which which binds to nuclear, or directly where the protein acts as autonomous nuclear transport receptor.
Import of macromolecules containing a NLS requires a heterodimer of importin alpha and beta subunits also called as karyopherins. Importin alpha binds the cytoplasmic NLS-containing protein and importin beta docks the complex at the cytoplasmic side of the nuclear pore complex. Movement through the nuclear pore is mediated by the small GTP binding protein Ran via an energy requiring mechanism. Using nucleotide triphosphates and Ran, the nuclear transport complex moves into the nuclear pore and the importin subunits dissociate. Importin alpha and its passenger protein enter the nucleoplasm while importin beta remains at the pore.

Specificity

This antibody recognizes Importin beta-1.

Immunogen

Epitope: Unknown
Recombinant protein corresponding to human Importin beta-1.

Application

Research Category
Epigenetics & Nuclear Function
Research Sub Category
RNA Metabolism & Binding Proteins
Use Anti-Importin beta-1 Antibody, clone 3H14 (Mouse Monoclonal Antibody) validated in WB, ICC to detect Importin beta-1 also known as Karyopherin subunit beta-1, Nuclear factor P97.

Quality

Evaluated by Western Blot in HeLa cell lysate.

Western Blot Analysis: 0.5 µg/ml of this antibody detected Importin beta-1 on 10 µg of HeLa cell lysate.

Target description

~ 97 kDa

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4, 150 mM NaCl) with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service