Documents
CLS9099
Corning® aspiration pipettes
volume 5 mL
Synonym(s):
Aspiration Pipette
About This Item
UNSPSC Code:
41121500
eCl@ss:
32070201
NACRES:
NB.01
material
polystyrene
sterility
sterile
packaging
case of 200 ea
pack of 50 ea
manufacturer/tradename
Corning 9099
capacity
5 mL
volume
5 mL
General description
  • Corning® aspirating pipettes are made of optically clearpolystyrene for durability and good mechanical strength.
  • The sterile,ungraduated, and unplugged pipettes are best suited for aspirating liquidaseptically using vacuum suction.
  • Individually wrappedand bulk packed.
  • Disposable andcertified non-pyrogenic.
Aspirating pipettes are sterile, ungraduated and unplugged polystyrene pipettes for aspirating liquid using vacuum suction.
Legal Information
Corning is a registered trademark of Corning, Inc.