Key Documents
CLS9016
Corning® aspiration pipettes
volume 2 mL, polystyrene, plain, paper/plastic wrapped
Synonym(s):
Aspiration Pipettes, Corning Pipettes
Select a Size
500 EA
€326.00
€326.00
Check Cart for Availability
Select a Size
Change View
500 EA
€326.00
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
€326.00
Check Cart for Availability
material
polystyrene
sterility
sterile
packaging
pack of 50 ea
case of 500 ea
manufacturer/tradename
Corning 9016
capacity
2 mL
volume
2 mL
General description
  • Corning® aspirating pipettes are made of optically clearpolystyrene for durability and good mechanical strength.
  • The sterile,ungraduated, and unplugged pipettes are best suited for aspirating liquidaseptically using vacuum suction.
  • Individually wrappedand bulk packed.
  • Disposable andcertified non-pyrogenic.
Aspirating pipettes are sterile, ungraduated and unplugged polystyrene pipettes for aspirating liquid using vacuum suction.
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)