Skip to Content
Merck
All Photos(1)

Documents

CLS432004

Corning® CoolCell LX Cell Freezing Container

(12 x 1 mL or 2 mL cryogenic vials), pink

Synonym(s):

cryovials

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41123300
NACRES:
NC.01

material

foamed PE

feature

Holds 12 x 1-2 mL cryovials

packaging

pkg of 1

manufacturer/tradename

Corning 432004

technique(s)

cryopreservation: suitable

H × diam.

10.11 cm × 11.7 cm , 12.7 mm well diameter

color

pink

capacity

(12 x 1 mL or 2 mL cryogenic vials)

Looking for similar products? Visit Product Comparison Guide

General description

Corning CoolCell LX, in combination with a -80°C freezer, will provide alcohol-free freezing at the rate of -1°C/minute that is ideal for cryopreservation of most cells and cell lines. Using a combination of uniform-density cross-linked polyethylene foam, a solid state core, and radial vial symmetry, freezing profiles are consistent and reproducible. The foam is non-absorbent and will impose negligible change in the freezer environment; thereby protecting nearby frozen samples. The low heat content also ensures that CoolCell LX containers will rapidly return to room temperature when removed from freezer.

  • For 12 standard 1.0 mL to 2.0 mL cryogenic vials.
  • Radially symmetric for uniform vial freezing.
  • Numbered wells for easy sample identification.
  • Beveled lid for secure gripping and easy opening.
  • Exposed vial tops when lid is open for quick, organized removal of frozen samples.

Legal Information

CoolCell is a trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service