CLS431225
Corning® syringe filters
Nylon membrane, pore size 0.45 μm
Synonym(s):
disposable syringe filters, sterile syringe filters, syringe filters
All Photos(1)
About This Item
Recommended Products
material
Nylon membrane
polystyrene housing
sterility
sterile
packaging
case of 50
manufacturer/tradename
Corning 431225
diam.
25 mm
pore size
0.45 μm
fitting
female Luer inlet (lock)
male Luer outlet (slip)
Related Categories
General description
Corning syringe filters are 100% integrity tested and are certified nonpyrogenic and noncytotoxic and are manufacturered in accordance with ISO 9002 standards. A variety of membranes are available to meet your needs.
PES for low protein binding and faster flow rates; surfactant-free cellulose acetate (SFCA) for the lowest protein binding; PTFE for chemical resistance; and regenerated cellulose (RC) the best choice for DMSO compatibility.
PES for low protein binding and faster flow rates; surfactant-free cellulose acetate (SFCA) for the lowest protein binding; PTFE for chemical resistance; and regenerated cellulose (RC) the best choice for DMSO compatibility.
These Corning syringe filters have polypropylene copolymer housings, female luer lock inlets and male luer slip outlets.
- Nylon (NY) membranes are naturally hydrophilic, surfactant-free and offer the lowest extractables
- 100% integrity tested, sterile and certified nonpyrogenic and noncytotoxic
- Manufactured in accordance with ISO 9002 standards
Legal Information
Corning is a registered trademark of Corning, Inc.