Documents
CLS431220
Corning® syringe filters
cellulose acetate membrane (surfactant-free), diam. 28 mm, pore size 0.45 μm
Synonym(s):
disposable syringe filters, sterile syringe filters, syringe filters
About This Item
UNSPSC Code:
41104922
NACRES:
NB.24
material
acrylic copolymer housing
cellulose acetate membrane (surfactant-free)
sterility
sterile
packaging
case of 50
manufacturer/tradename
Corning 431220
diam.
28 mm
pore size
0.45 μm
fitting
female Luer inlet (lock)
male Luer outlet (slip)
Related Categories
General description
Corning syringe filters are 100% integrity tested and are certified nonpyrogenic and noncytotoxic and are manufacturered in accordance with ISO 9002 standards. A variety of membranes are available to meet your needs.
PES for low protein binding and faster flow rates; surfactant-free cellulose acetate (SFCA) for the lowest protein binding; PTFE for chemical resistance; and regenerated cellulose (RC) the best choice for DMSO compatibility.
These Corning syringe filters have acrylic copolymer housings, female luer lock inlets and male luer slip outlets.
  • SFCA-PF (surfactant-free cellulose acetate with prefilter) membranes provide the lowest protein binding and have an integral glass fiber prefilter for extra filtering capacity
  • 100% integrity tested, sterile and certified nonpyrogenic and noncytotoxic
  • Manufactured in accordance with ISO 9002 standards
Legal Information
Corning is a registered trademark of Corning, Inc.
Visit the Document Library
Qiushi Wei et al.
International journal of molecular medicine, 45(3), 816-824 (2020-01-28)
Icaritin, a metabolite of icariin, is a potent promoter of bone marrow‑derived mesenchymal stem cells (BMSCs) osteogenesis, but the underlying mechanisms remain unclear. To examine the effects of icaritin on osteogenic differentiation, BMSCs were exposed to osteogenic induction medium with