CLS3727
Corning® 1536 well plates
flat bottom white polystyrene wells, NBS™ (TC-treated), 50/cs, sterile, lid
Synonym(s):
1536 multiwell plate, 1536 well microplate, 1536 well microtiter plate, 1536 well plate, HTS plate, TC-treated plate, immunoassay plate, tissue-culture treated plate
About This Item
Recommended Products
material
flat bottom white polystyrene wells
polystyrene plate
white plate
sterility
sterile
feature
lid
skirt
packaging
pack of 10
case of 50
manufacturer/tradename
Corning 3727
maximum volume
12.5 μL
size
1536 wells
surface area
0.03 cm2
working volume
8 μL
color
white
suitability
suitable for (fluorescent; luminescent assays)
binding type
Tissue Culture (TC)-treated surface
Looking for similar products? Visit Product Comparison Guide
General description
- Excellent signal dynamic range due to low background fluorescence and enhanced signal intensity
- Superior flatness minimizes variability within a plate and between plates
- Flat bottom wells are compatible with automated dispensers
- NBS™ microplates are ideal for homogeneous assays with "sticky" enzymes or for reducing nonspecific binding to microplate wells
- Working volume of up to 10 μL
- 1536 well polystyrene plates
- Low background fluorescence, minimal light scatter and reduced crosstalk
- Excellent signal dynamic range due to low background fluorescence and enhanced signal intensity
- Opaque walls to prevent well-to-well crosstalk
- Flat well bottoms possess superior flatness -minimizes variability within a plate and between plates
- Round well bottoms for reduced air entrapment and improved CV′s and Z factor
- Raised well bottoms for high sensitivity
- Flood reservior on four sides to reduce instrument contamination
- Suitable for fluorescent and luminescent assays
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service