Skip to Content
Merck
All Photos(1)

Documents

CLS3891

Corning® 1536 well plates

flat bottom clear polystyrene wells, non-treated, 50/cs, non-sterile, lid: no

Synonym(s):

1536 multiwell plate, 1536 well microplate, 1536 well microtiter plate, 1536 well plate, HTS plate, TC-treated plate, immunoassay plate, tissue-culture treated plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.24

material

black plate
flat bottom clear polystyrene wells
polystyrene plate

sterility

non-sterile

feature

lid: no
skirt

packaging

pack of 10
case of 50

manufacturer/tradename

Corning 3891

size

1536 wells

well volume

12.5 μL

well working volume

8 μL

color

clear

suitability

suitable for (fluorescent; luminescent assays)

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

Corning 1536 Well Plates

  • 1536 well polystyrene plates
  • Low background fluorescence, minimal light scatter and reduced crosstalk
  • Excellent signal dynamic range due to low background fluorescence and enhanced signal intensity
  • Opaque walls to prevent well-to-well crosstalk
  • Flat well bottoms possess superior flatness -minimizes variability within a plate and between plates
  • Round well bottoms for reduced air entrapment and improved CV′s and Z factor
  • Raised well bottoms for high sensitivity
  • Flood reservior on four sides to reduce instrument contamination
  • Suitable for fluorescent and luminescent assays
  • Designed for fluorescent homogeneous assays
  • Superior performance compared to competitor plates: Lower CVs, higher signal-to-noise ratios, and Lower background fluorescence
  • Processed for superior flatness with minimal variability within plate and between plates

Legal Information

Corning is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service