Skip to Content
Merck

Skip To

C9745

Anti-Cullin 3 (C-terminal) antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-CUL3

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IF, IP, WB
Citations:
1


biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~80 kDa

species reactivity

rat, human, mouse

concentration

~1.0 mg/mL

technique(s)

immunoprecipitation (IP): 1-2 μg using lysate of rat brain, indirect immunofluorescence: 2-5 μg/mL using mouse 3T3 cells, western blot: 2-5 μg/mL using whole extract of human Jurkat cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CUL3(8452)
mouse ... Cul3(26554)
rat ... Cul3(301555)

Biochem/physiol Actions

Anti-Cullin 3 (C-terminal) recognizes human, mouse, and rat Cullin 3.

Physical form

Solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide.

Preparation Note

For continuous use, store at 2–8 °C for up to one month. For extended storage, freeze in working aliquots at –20 °C. Repeated freezing and thawing is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)

wgk

nwg

Storage Class

12 - Non Combustible Liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

Instructions


M T M van Jaarsveld et al.
Oncogene, 32(36), 4284-4293 (2012-10-10)
Epithelial ovarian cancer is the most lethal gynecological malignancy in the Western world. A major impediment for the successful treatment is the development of drug resistance. The molecular processes that contribute to resistance have been extensively studied; however, there is



Global Trade Item Number

SKUGTIN
C9745-200UL04061838030573

Questions

Reviews

No rating value

Active Filters