Skip to Content
Merck

Skip To

C8743

Cholesterol Dehydrogenase from Nocardia sp.

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
500 units
Check Cart for Availability
€ 264,00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54
EC Number:
1.1.1.B40
Specific activity:
≥18 unit/mg solid
Recombinant:
expressed in Streptomyces lividans

€ 264,00


Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


recombinant

expressed in Streptomyces lividans

Quality Segment

form

lyophilized solid

specific activity

≥18 unit/mg solid

mol wt

~37 kDa by SDS-PAGE

storage temp.

−20°C

Application

Enzyme used in determining cholesterol in serum
The enzyme sequence is derived from Nocardia sp., and this cholesterol dehydrogenase is expressed in Streptomyces lividans. Cholesterol Dehydrogenase from Nocardia sp. has been used in the preparation of amperometric cholesterol biosensor for cholesterol detection in biological samples and in a study to investigate the atomic structure of the autosomal recessive hypercholesterolemia phosphotyrosine-binding domain with the LDL-receptor tail. Cholesterol dehydrogenase from Nocardia sp. Has also been used in a study to assess organization, dynamics, and segregation of Ras nanoclusters in membrane domains.

Biochem/physiol Actions

Cholesterol dehydrogenase is a NAD(P) dependent microbial dehydrogenase that oxidizes the 3-β-hydroxyl group of cholesterol to yield cholest-4-ene-3-one. Molecular mass of the protein is found to be 37 kDa based on SDS-PAGE. Its pH is 4.5.
Cholesterol dehydrogenase (CDH) is a nicotinamide adenine dinucleotide phosphate (NAD(P)) dependent microbial dehydrogenase that oxidizes the 3-β-hydroxyl group of cholesterol to yield cholest-4-ene-3-one. Cholesterol detection by using CDH allows direct and easier quantification compared to cholesterol oxidase based assay.

Physical form

Supplied as a lyophilized powder

Other Notes

One unit produces 1.0 μmole of cholesten-4-en-3-one per minute at pH 8.5 at 25 °C.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C3766C4405S0937
specific activity

≥18 unit/mg solid

specific activity

≥200 units/g protein

specific activity

8-20 units/mg solid

specific activity

≥45 units/mg solid

recombinant

expressed in Streptomyces lividans

recombinant

-

recombinant

-

recombinant

expressed in E. coli

form

lyophilized solid

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

mol wt

~37 kDa by SDS-PAGE

mol wt

84 kDa by SDS-PAGE

mol wt

71 kDa by SDS-PAGE, 83 kDa by gel filtration

mol wt

56 kDa by SDS-PAGE

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters