Skip to Content
Merck

Skip To

C7290

N-Z-Amine® A

suitable for plant cell culture

Synonym(s):

Peptone from casein

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
250 g
Check Cart for Availability
€ 75,50
500 g
Check Cart for Availability
€ 134,00
1 kg
Check Cart for Availability
€ 237,00

About This Item

CAS Number:
MDL number:
NACRES:
NA.72
UNSPSC Code:
10171502
Form:
powder

€ 75,50


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

powder

Quality Segment

nitrogen analysis

~6.5% amino, ~13.1% total

technique(s)

cell culture | plant: suitable

application(s)

agriculture

General description

N-Z-Amine A is a peptone containing amino acids and peptides derived by the enzymatic hydrolysis of casein protein. N-Z-Amine A may be used as a plant media growth supplement and in microbiological work.

Application

N-Z-Amine® A has been used:
  • as a component in MS media add this for in in vitro shoot multiplication
  • as a component in MS media add this for callus induction
  • as a component in N6-basal medium for immature zygotic embryos culturing

Legal Information

N-Z-Amine is a registered trademark of Kerry Group

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C0626N4517C7210
technique(s)

cell culture | plant: suitable

technique(s)

-

technique(s)

-

technique(s)

microbiological culture: suitable

form

powder

form

powder

form

powder

form

powder

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

100

application(s)

agriculture

application(s)

food and beverages
microbiology

application(s)

microbiology

application(s)

-

nitrogen analysis

~6.5% amino, ~13.1% total

nitrogen analysis

≥6% amino, ≥11.0% total

nitrogen analysis

≥6% amino, ≥11% total

nitrogen analysis

-


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters