Merck
Skip To
C6557
Nunc® Lab-Tek® II Chamber Slide system
wells 2, well area 4 cm2, sterile
Synonym(s):
cell culture chamber, chamber slides, microscope slides, microscopy slides, Nunc 154461
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Size/SKUAvailabilityPrice
16 ea
Check Cart for Availability
€ 352,00
96 ea
Check Cart for Availability
€ 1.190,00
About This Item
NACRES:
NB.22
UNSPSC Code:
41122600
Material:
colorless , polystyrene chamber
Sterility:
sterile
Packaging:
pack of 16 ea, case of 96 ea
Wells:
2
€ 352,00
Check Cart for Availability
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
colorless , polystyrene chamber
sterility
sterile
feature
CE marked
packaging
pack of 16 ea, case of 96 ea
manufacturer/tradename
Nunc 154461
well area
4 cm2
wells
2
working volume
1.0-2.0 mL
suitability
suitable for (cell culture; attachment and growth of cells)
General description
Grow, stain, and examine cells directly on the Nunc® Lab-Tek® II chamber slide which consists of a removable polystyrene media chamber attached to a standard glass slide. Slide surface is specially treated for adherent cell culture. Available in 1-, 2-, 4-, and 8-well formats.
Features and Benefits
  • Slide surface is specially treated with a unique RS wash to ensure consistent cell growth.
  • Tabbed polystyrene lid allows gas exchange with minimal evaporation.
  • Removal tool separates media chamber from glass slide.
  • Biocompatible adhesive hydrophobic well border is printed on slide.
  • Slide features a white frosted writing area.
  • Includes: Chamber removal tool.
  • Sterile
  • Packed in convenient stackable incubation trays.
Legal Information
Chamber Slide is a trademark of Thermo Fisher Scientific or its subsidiaries
Lab-Tek is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Compare Similar Items
View Full Comparison
Show Differences
1 of 1
This Item
C6307C6807C7057
material
colorless , polystyrene chamber
material
colorless , polystyrene chamber
material
colorless , polystyrene chamber
material
polystyrene chamber
manufacturer/tradename
Nunc 154461
manufacturer/tradename
Nunc 154453
manufacturer/tradename
Nunc 154526
manufacturer/tradename
Nunc 154534
sterility
sterile
sterility
sterile
sterility
sterile
sterility
sterile
wells
2
wells
1
wells
4
wells
8
packaging
pack of 16 ea, case of 96 ea
packaging
pack of 16 ea, case of 96 ea
packaging
pack of 16 ea, case of 96 ea
packaging
pack of 16 ×
feature
CE marked
feature
CE marked
feature
CE marked
feature
CE marked
Still not finding the right product?
Choose from one of the most recent versions:
Certificates of Analysis (COA)