Skip to Content
Merck

Skip To

C5024

Carbonic Anhydrase from bovine erythrocytes

non-denaturing PAGE marker

Synonym(s):

Carbonate Dehydratase, Carbonate Hydrolyase

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 vial
Check Cart for Availability
€ 131,00

About This Item

CAS Number:
UNSPSC Code:
12352204
NACRES:
NA.54
EC Number:
232-576-6
MDL number:
EC Number:

€ 131,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

lyophilized powder

mol wt

~29 kDa

suitability

suitable for marker for electrophoresis (non-denaturing PAGE)

storage temp.

−20°C

Application

This product is used as a marker to determine the molecular mass of an unknown protein in non-denaturing electrophoresis systems. In such systems, proteins often retain their biological activity, thus allowing in-gel detection using specific substrate stains. This technique also facilitates the recovery of the active protein. The product yields 3 bands.

Preparation Note

0.9-1.4 mg/mL after reconstitution with 1 mL of water

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C2273C3666C2624
form

lyophilized powder

form

powder

form

powder

form

lyophilized powder

suitability

suitable for marker for electrophoresis (non-denaturing PAGE)

suitability

-

suitability

-

suitability

-

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

2-8°C

mol wt

~29 kDa

mol wt

~29 kDa

mol wt

-

mol wt

-

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200


Still not finding the right product?


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters