Skip to Content
Merck

Skip To

C1495

CSK, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
10 μg
Check Cart for Availability
€ 385,00

About This Item

NACRES:
NA.32
UNSPSC Code:
51111800
Specific activity:
361-489 nmol/min·mg
Assay:
≥70% (SDS-PAGE)
Recombinant:
expressed in baculovirus infected Sf9 cells

€ 385,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


recombinant

expressed in baculovirus infected Sf9 cells

Quality Segment

product line

PRECISIO® Kinase

assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

361-489 nmol/min·mg

mol wt

~78 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CSK(1445)

Biochem/physiol Actions

CSK is a cytoplasmic tyrosine kinase that has been shown to downregulate the tyrosine kinase activity of the c-src through tyrosine phosphorylation of the c-src carboxy terminus. A yeast 2-hybrid system has been used to identify proteins associated with CSK. The Src homology-3 (SH3) domain of CSK associates with a proline-rich region of PEP, a protein-tyrosine phosphatase expressed in hemopoietic cells. This association is highly specific and it is speculated that PEP may be an effector and/or regulator of CSK in T cells and other hemopoietic cells.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
K3518K4268SRP5016
assay

≥70% (SDS-PAGE)

assay

≥70% (SDS-PAGE)

assay

≥70% (SDS-PAGE)

assay

≥70% (SDS-PAGE)

Gene Information

human ... CSK(1445)

Gene Information

human ... PIM2(11040)

Gene Information

human ... PRKD1(5587)

Gene Information

human ... CSNK1G2(1455)

specific activity

361-489 nmol/min·mg

specific activity

259-351 nmol/min·mg

specific activity

578-782 nmol/min·mg

specific activity

25-35 nmol/min·mg

recombinant

expressed in baculovirus infected Sf9 cells

recombinant

expressed in baculovirus infected Sf9 cells

recombinant

expressed in baculovirus infected Sf9 cells

recombinant

expressed in baculovirus infected Sf9 cells

form

buffered aqueous glycerol solution

form

buffered aqueous glycerol solution

form

buffered aqueous glycerol solution

form

buffered aqueous glycerol solution

mol wt

~78 kDa

mol wt

~61 kDa

mol wt

~131 kDa

mol wt

~73 kDa


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Global Trade Item Number

SKUGTIN
C1495-10UG04061837753329

Questions

Reviews

No rating value

Active Filters