Skip to Content
Merck

Skip To

C-14045

Normal Human Epidermal Melanocytes 3 (NHEM 3)

Adult, isolated in PMA-free Melanocyte Growth Medium M3, 1 million cells per pellet

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NA.81
UNSPSC Code:
41106514
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−20°C

General description

Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details. Cells are dissolved in RNAlater for subsequent RNA, DNA, or protein analysis. Note that cell pellets cannot be revived.

Application

Primary Normal Human Epidermal Melanocytes (NHEM) are isolated from the epidermis of adult skin (different locations) and are provided in a proliferating format. Melanocytes are located in the stratum basale, but branch out between the keratinocytes in suprabasal layers. About 5-10 % of the cells in the epidermis are melanocytes. The main purpose of melanocytes is the production of melanin, the protein responsible for the pigmentation of the skin, eyes, and hair. Melanin protects the cells in the skin and in deeper layers from the hazardous effects of UV radiation. It is produced and stored in melanosomes, which are located close to the melanocyte membrane, but it can also be transferred to neighboring keratinocytes.
Normal Human Epidermal Melanocytes M3 are isolated using the serum-free, BPE-free and PMA-free Melanocyte Growth Medium M3. Since PMA is a tumor promoting mitogen, it can interfere with experimental approaches. Therefore, we recommend using cells isolated in Melanocyte Growth Medium M3 and also using this medium for cultivation. Normal Human Epidermal Keratinocytes (NHEK) and Normal Human Dermal Fibroblasts (NHDF) from the same donor are available on request.

Preparation Note

Click here for more information.

Analysis Note

Rigid quality control tests are performed for each lot of the corresponding cells. They are tested for cell viability, cell morphology, growth performance and for cell-type specific markers. In addition, all cells have been tested for the absence of HIV-1, HIV-2, HBV, HCV, HTLV-1, HTLV-2 and microbial contaminants (fungi, bacteria, and mycoplasma).

Other Notes

The Cell Pellets can be stored indefinitely at -20°C, and for up to 1 month at 4°C. Note: The Cell Pellets will not freeze at -20°C. Always centrifuge product briefly before opening vial.

Disclaimer

Although tested negative for HIV-1, HIV-2, HBV, HCV, HTLV-1 and HTLV-2, the cells – like all products of human origin – should be handled as potentially infectious. No test procedure can completely guarantee the absence of infectious agents.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C-12413C-12422C-12463
technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

shipped in

ambient

storage temp.

−20°C

storage temp.

−196°C

storage temp.

−196°C

storage temp.

no temp limit


Still not finding the right product?


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

7783-20-2

CAS No.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
C-1404504065268043660

Questions

Reviews

No rating value

Active Filters