Select a Size
| Pack Size | SKU | Availability | Price |
|---|---|---|---|
| 1 ea | Check Cart for Availability | € 1.570,00 |
About This Item
biological source
human adipose tissue
packaging
pkg of 500,000 cells
morphology
(stromal)
technique(s)
cell culture | mammalian: suitable
shipped in
ambient
storage temp.
room temp
General description
Application
Biochem/physiol Actions
Preparation Note
Analysis Note
Other Notes
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| biological source human adipose tissue | biological source human umbilical cord matrix (Wharton′s jelly) | biological source human umbilical cord matrix (Wharton′s jelly) | biological source human umbilical cord matrix (Wharton′s jelly) |
| technique(s) cell culture | mammalian: suitable | technique(s) cell culture | mammalian: suitable | technique(s) - | technique(s) cell culture | mammalian: suitable |
| morphology (stromal) | morphology (stromal) | morphology (stromal) | morphology (stromal) |
| packaging pkg of 500,000 cells | packaging pkg of 500,000 cells | packaging pkg of 1,000,000 cells | packaging pkg of 500,000 cells |
| storage temp. room temp | storage temp. −196°C | storage temp. −20°C | storage temp. room temp |
| shipped in ambient | shipped in dry ice | shipped in dry ice | shipped in ambient |
Still not finding the right product?
Explore all of our products under Human Mesenchymal Stem Cells (hMSC)
Storage Class
12 - Non Combustible Liquids
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
10108-64-2
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Protocols
Information about mesenchyme, specifically mesenchymal stem cell procotols. Step-by-step cell culture protocols for mesenchymal stem cell (MSC) isolation, expansion and differentiation.
Articles
Are your mesenchymal stem cells really MSCs? Learn more about the MSC characterization guidelines proposed by the International Society for Cellular Therapy (ISCT).
Frequently asked questions about mesenchymal stem cells including MSC derivation, expansion, differentiation and allogenic stem cell therapy.
Protocol to differentiate human mesenchymal stem cells into neurons with extensive dendrites and axons in 3 days. Explore over 350 PromoCell products on Sigma.com.

