Select a Size
| Pack Size | SKU | Availability | Price |
|---|
About This Item
biological source
human foreskin (juvenile epidermis)
form
aqueous solution
packaging
pkg of 500,000 cells
morphology
( epidermal)
technique(s)
cell culture | mammalian: suitable
shipped in
ambient
storage temp.
room temp
General description
Application
Biochem/physiol Actions
Preparation Note
Analysis Note
Other Notes
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| technique(s) cell culture | mammalian: suitable | technique(s) - | technique(s) - | technique(s) cell culture | mammalian: suitable |
| biological source human foreskin (juvenile epidermis) | biological source human foreskin (juvenile epidermis) | biological source human epidermis (adult) | biological source human epidermis |
| morphology ( epidermal) | morphology (epidermal) | morphology (epidermal) | morphology ( epidermal) |
| shipped in ambient | shipped in dry ice | shipped in dry ice | shipped in ambient |
| storage temp. room temp | storage temp. −20°C | storage temp. −20°C | storage temp. room temp |
| packaging pkg of 500,000 cells | packaging pkg of 1,000,000 cells | packaging pkg of 1,000,000 cells | packaging pkg of 500,000 cells |
Still not finding the right product?
Explore all of our products under Normal Human Epidermal Keratinocytes (NHEK)
Storage Class
12 - Non Combustible Liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
7791-20-0
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Primary human keratinocytes and serum-free keratinocyte growth media for the feeder cell-free in vitro cultivation of epidermal keratinocytes. Protocols for handling, cell passaging, media, and product use.

