C112080
DMT-dC(tac) Phosphoramidite
configured for PerkinElmer, configured for Polygen
Synonym(s):
DMT-dC(tac) amidite, N-[2-[4-(1,1-dimethylethyl)phenoxy]acetyl]-5′-O-[bis(4-methoxyphenyl)phenylmethyl]-2′-deoxycytidine, 3′-[2-cyanoethyl N,N-bis(1-methylethyl)phosphoramidite]
Select a Size
About This Item
Recommended Products
type
for DNA synthesis
Quality Level
product line
Proligo Reagents
Assay
≥98% (31P-NMR)
≥98.0% (reversed phase HPLC)
form
solid
technique(s)
oligo synthesis: suitable
λ
conforms (UV/VIS Identity)
compatibility
configured for PerkinElmer
configured for Polygen
nucleoside profile
base: deoxycytidine
base protecting group: TAC
2' protecting group: none
5' protecting group: DMT
deprotection: fast
storage temp.
-10 to -25°C
SMILES string
CC(C)N(C(C)C)P(OCCC#N)O[C@H]1C[C@@H](O[C@@H]1COC(c2ccccc2)(c3ccccc3)c4ccccc4)N5C=CC(NC(=O)COc6ccc(cc6)C(C)(C)C)=NC5=O
InChI
1S/C49H58N5O7P/c1-35(2)54(36(3)4)62(59-31-17-29-50)61-42-32-46(53-30-28-44(52-47(53)56)51-45(55)34-57-41-26-24-37(25-27-41)48(5,6)7)60-43(42)33-58-49(38-18-11-8-12-19-38,39-20-13-9-14-21-39)40-22-15-10-16-23-40/h8-16,18-28,30,35-36,42-43,46H,17,31-34H2,1-7H3,(H,51,52,55,56)/t42-,43+,46+,62?/m0/s1
InChI key
NHKNJNORWFDRBB-ROEGVEHFSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service