Skip to Content
Merck

Skip To

T401000

DMT-dT-CPG

pore size 1000 Å, 25-35μmol/g

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 g
Check Cart for Availability
€ 45,20
10 g
Check Cart for Availability
€ 359,00

About This Item

NACRES:
NA.51
UNSPSC Code:
12352200
Assay:
≥98.0% (Nucleoside Purity, reversed phase HPLC)
Storage temp.:
2-8°C

€ 45,20


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

type

for DNA synthesis

product line

Proligo Reagents

assay

≥98.0% (Nucleoside Purity, reversed phase HPLC)

pore size

1000 Å

capacity

≥99 % (Coupling Efficiency)

λ

conforms (UV/VIS Identity)

storage temp.

2-8°C


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
G702000U701000C702000
description

pore size 1000 Å, 25-35μmol/g

description

pore size 1000 Å, 20-30 μmol/g

description

pore size 1000 Å, 20-30 μmol/g

description

pore size 1000 Å, 20-30 μmol/g

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

assay

≥98.0% (Nucleoside Purity, reversed phase HPLC)

assay

≥98.0% (Nucleoside Purity, reversed phase HPLC)

assay

≥98.0% (Nucleoside Purity, reversed phase HPLC)

assay

≥98.0% (Nucleoside Purity, reversed phase HPLC)

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

capacity

≥99 % (Coupling Efficiency)

capacity

≥98 % coupling capacity (Coupling Efficiency)

capacity

≥98 % coupling capacity (Coupling Efficiency)

capacity

≥98 % coupling capacity (Coupling Efficiency)

product line

Proligo Reagents

product line

-

product line

-

product line

-

type

for DNA synthesis

type

-

type

-

type

-


Still not finding the right product?

Explore all of our products under DMT-dT-CPG


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters