Skip to Content
Merck

C702000

DMT-rC(tac)-CPG

pore size 1000 Å, 20-30 μmol/g

Sign In to View Organizational & Contract Pricing.

Select a Size

C702000-1G

€ 256,00

€ 256,00


Check Cart for Availability

Request a Bulk Order

About This Item

NACRES:
NA.51
UNSPSC Code:
12352200
Assay:
≥98.0% (Nucleoside Purity, reversed phase HPLC)
Storage temp.:
−20°C

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Quality Level

assay

≥98.0% (Nucleoside Purity, reversed phase HPLC)

pore size

1000 Å

capacity

≥98 % coupling capacity (Coupling Efficiency)

storage temp.

−20°C

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
A702000G702000U701000
Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

assay

≥98.0% (Nucleoside Purity, reversed phase HPLC)

assay

≥98.0% (Nucleoside Purity, reversed phase HPLC)

assay

≥98.0% (Nucleoside Purity, reversed phase HPLC)

assay

≥98.0% (Nucleoside Purity, reversed phase HPLC)

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

pore size

1000 Å

pore size

1000 Å

pore size

1000 Å

pore size

1000 Å

capacity

≥98 % coupling capacity (Coupling Efficiency)

capacity

≥98 % coupling capacity (Coupling Efficiency)

capacity

≥98 % coupling capacity (Coupling Efficiency)

capacity

≥98 % coupling capacity (Coupling Efficiency)

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service