Skip to Content
Merck

Skip To

C1099

Caspase 8 human

≥90% (SDS-PAGE), recombinant, expressed in E. coli (C-terminal histidine tagged), buffered aqueous solution, ≥1,000 units/mg protein

Synonym(s):

FLICE, MASH, Mch5

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12352204
NACRES:
NA.32
MDL number:


recombinant

expressed in E. coli (C-terminal histidine tagged)

Quality Segment

assay

≥90% (SDS-PAGE)

form

buffered aqueous solution

specific activity

≥1,000 units/mg protein

mol wt

~30 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CASP8(841)

General description

Human recombinant C−terminal histidine tagged caspase 8 is expressed as the truncated pro-caspase 8 (amino acids 213−496). It is a fully active protein consisting of 18 kDa and 12.4 kDa subunits; the 12.4 kDa subunit contains the histidine tag.

Biochem/physiol Actions

Caspase 8 is at the top of the hierarchy of the caspase cascade and is considered an initiator caspase.
Caspase 8 is at the top of the hierarchy of the caspase cascade and is considered an initiator caspase. It exists in the cell as an inactive 55 kDa proenzyme. It is converted to the active form, consisting of 18 kDa and 11 kDa subunits, upon its recruitment to the cytoplasmic domain of activated death receptors such as CD95 or TNFR via the adapter protein FADD. The activation of the proenzyme is triggered by the protein′s aggregation, which leads to auto- or transprocessing. Caspase 8 activates downstream caspases (caspases 3, 6, and 7) that cleave key cellular substrates and lead to apoptotic death of the cells.

Physical form

Solution in 10% sucrose containing 20 mM Tris-HCl, 150 mM imidazole-HCl, pH 8.0, 20 mM 2-mercaptoethanol, 500 mM NaCl, 2.5 mM EDTA, and 0.1% CHAPS.

Other Notes

One unit will cleave 1.0 nmol of Ac-Ile-Glu-Thr-Asp-pNA per min at pH 7.4 at 25 °C.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C1224SAB5700197SCP0094
assay

≥90% (SDS-PAGE)

assay

≥85% (SDS-PAGE)

assay

-

assay

≥95% (HPLC)

recombinant

expressed in E. coli (C-terminal histidine tagged)

recombinant

expressed in E. coli (C-terminal histidine-tagged)

recombinant

-

recombinant

-

Quality Level

200

Quality Level

200

Quality Level

100

Quality Level

-

form

buffered aqueous solution

form

buffered aqueous glycerol solution

form

buffered aqueous solution

form

lyophilized

shipped in

dry ice

shipped in

dry ice

shipped in

wet ice

shipped in

-

UniProt accession no.

Q14790

UniProt accession no.

P42574

UniProt accession no.

Q14790

UniProt accession no.

-


pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - Skin Sens. 1

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

288-32-4

CAS No.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters