Skip to Content
Merck
All Photos(1)

Key Documents

BMSIPD1196MP

Sigma-Aldrich

Toughened Genomics 96 Deep Well Plate for BeadBlaster 96

polyethylene, capacity 2 mL, pack of 1 ×

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000
NACRES:
NB.15
Pricing and availability is not currently available.

material

polyethylene

manufacturer/tradename

Benchmark Scientific IPD1196-MP

packaging

pack of 1 ×

capacity

2 mL

compatibility

BeadBlaster 96

General description

Toughened polyethylene 96 square well genomics plate. 2.0 mL per well.

Application

genomics

Features and Benefits

  • Toughened
  • Deep well

Legal Information

BeadBlaster is a trademark of Benchmark Scientific, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service