AXYP2MLSQCS
Deep well plate
25 ea of, sterile
Synonym(s):
Deep well plate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
square bottom clear polypropylene wells
sterility
sterile
feature
lid: no
skirt
packaging
case of 25 ea
pack of 25 ea
25 ea of
manufacturer/tradename
Axygen Inc P-2ML-SQ-C-S
Corning P2MLSQCS
maximum volume
2.2 mL
size
96 wells
binding type
poly-D-lysine coated surface
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Deep well plate has been used for:
- long-term storage
- collecting sample
- In vitro growth chambers
- flow cytometry assays
Features and Benefits
- Provides proper heat-sealing film due to ultra-flat surface
- Endure temperatures as low as -80°C and highly chemical resistant
- Made up of polypropylene
Legal Information
Corning is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Molecular cell, 81(24), 5099-5111 (2021-12-18)
The SARS-CoV-2 spike protein is a critical component of vaccines and a target for neutralizing monoclonal antibodies (nAbs). Spike is also undergoing immunogenic selection with variants that increase infectivity and partially escape convalescent plasma. Here, we describe Spike Display, a
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service