BAF189700001
Bel-Art® Tissue Culture Flask Rack
Holds 12 x 75 mL flasks, polypropylene
Select a Size
€110.00
Select a Size
About This Item
€110.00
Recommended Products
material
polypropylene
feature
Holds 12 x 75 mL flasks
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F18970-0001
well L
25 cm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Racks to hold either 25cm² or 75cm² (50-75ml or 250-260ml) or similar sized flasks in an upright position during processing of tissue cultures.
- Prevents "domino effect" of flasks tipping
- Helps organize flasks when collecting or changing medium, washing of cell monolayer, trypsinization and seeding of cells
- Grips on each end aid in handling and transport of flasks to the incubator
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service