Skip to Content
Merck
All Photos(1)

Key Documents

BAF187453017

No-Wire Multi-Tube Racks

No-Wire™ Multi-Tube Racks
1 of 1 reviewers received a sample product or took part in a promotion

Holds 6 of 50 mL and 12 of 15 mL Centrifuge Tubes

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€60.10

€60.10


Check Cart for Availability


Select a Size

Change View
1 EA
€60.10

About This Item

UNSPSC Code:
41122804
NACRES:
NB.22

€60.10


Check Cart for Availability

material

white polypropylene

feature

autoclavable
Holds 12 x 15 ml tubes
Holds 6 x 50 mL tubes

packaging

pack of 1 ea

manufacturer/tradename

Bel-Art F18745-3017

Looking for similar products? Visit Product Comparison Guide

General description

Each No-Wire Racks holds two or three different sizes of round or conical bottom centrifuge tubes. One rack holds four 50ml, six 15ml and twenty-five 1.5ml tubes (hole sizes, 30.2mm, 17.3mm and 11.4mm respectively), other rack holds six 50ml and twelve 15ml centrifuge tubes (holes size, 30.2mm and 17.3mm respectively). Easy to assemble. Dimensions are 246mm x 104mm x 64mm high (9.7" x 4.1" x 2.5"). The white polypropylene racks are submersible and steam autoclavable to 121ºC (250º F).
Store different size tubes of required reagents using one space saving rack
  • Ships flat - Easy to assemble

Legal Information

No-Wire is a trademark of SP Scienceware, Division of SP

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

1 of 1 reviewers received a sample product or took part in a promotion

Active Filters

  1. Montreal CANADA
    • Review 1
    • Votes 0
    5 out of 5 stars.

    Convenient multi tubes rack

    Appreciate that the rack was not taking a lot of space during shipping. But it's still robust, stays easily clean and having 50ml and 15ml tubes side by side on the same rack is extremely convenient when preparing solutions from stocks or having all the stock solutions in front of you.

    Helpful?

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service