Skip to Content
Merck

Skip To

B8791

5-Bromo-5-nitro-1,3-dioxane

≥99%

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
5 g
Check Cart for Availability
€ 195,00

About This Item

Empirical Formula (Hill Notation):
C4H6BrNO4
CAS Number:
Molecular Weight:
212.00
NACRES:
NA.21
PubChem Substance ID:
UNSPSC Code:
12352119
EC Number:
250-001-7
MDL number:

€ 195,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥99%

form

powder or crystals

antibiotic activity spectrum

Gram-negative bacteria, Gram-positive bacteria, fungi

application(s)

diagnostic assay manufacturing

mode of action

enzyme | inhibits

SMILES string

[O-][N+](=O)C1(Br)COCOC1

InChI

1S/C4H6BrNO4/c5-4(6(7)8)1-9-3-10-2-4/h1-3H2

InChI key

XVBRCOKDZVQYAY-UHFFFAOYSA-N

Application

  • Development of a solid-phase microextraction gas chromatography with microelectron-capture detection method for the determination of 5-bromo-5-nitro-1,3-dioxane in rinse-off cosmetics.: This article details the development of a sensitive analytical method for detecting 5-bromo-5-nitro-1,3-dioxane in cosmetics, enhancing our ability to monitor and ensure the safety of cosmetic products (Fernandez-Alvarez et al., 2010).

Disclaimer

The product is not intended for use as a biocide under global biocide regulations, including but not limited to US EPA′s Federal Insecticide Fungicide and Rodenticide Act, European Biocidal Products Regulation, Canada’s Pest Management Regulatory Agency, Turkey’s Biocidal Products Regulation, Korea’s Consumer Chemical Products and Biocide Safety Management Act (K-BPR) and others.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
393363429554429546
mode of action

enzyme | inhibits

mode of action

-

mode of action

-

mode of action

-

antibiotic activity spectrum

Gram-negative bacteria, fungi, Gram-positive bacteria

antibiotic activity spectrum

-

antibiotic activity spectrum

-

antibiotic activity spectrum

-

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

assay

≥99%

assay

99%

assay

99%

assay

99%

form

powder or crystals

form

powder or crystals

form

powder or crystals

form

powder or crystals

application(s)

diagnostic assay manufacturing

application(s)

diagnostic assay manufacturing
hematology
histology

application(s)

diagnostic assay manufacturing
environmental
hematology
histology

application(s)

diagnostic assay manufacturing
environmental
hematology
histology


Still not finding the right product?

Explore all of our products under 5-Bromo-5-nitro-1,3-dioxane


signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1A

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk

WGK 3

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
B8791-5G04061835542963

Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. What temperature should B8791 be stored at?

    1 answer
    1. This product may be stored at room temperature.

      Helpful?

Reviews

No rating value

Active Filters