Skip to Content
Merck

Skip To

B4062

BRAF (416-766), active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

BRAF1, MGC126806, MGC138284, RAFB1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 μg
Check Cart for Availability
€ 503,00

About This Item

NACRES:
NA.32
UNSPSC Code:
12352200
Specific activity:
63-85 nmol/min·mg
Assay:
≥70% (SDS-PAGE)
Recombinant:
expressed in baculovirus infected Sf9 cells

€ 503,00


Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


recombinant

expressed in baculovirus infected Sf9 cells

Quality Segment

product line

PRECISIO® Kinase

assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

63-85 nmol/min·mg

mol wt

~63 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... BRAF(673)

Biochem/physiol Actions

BRAF is a member of the RAF family that is activated by members of the Ras family upon growth factor-induced stimulation. Active Ras can induce heterodimerization of cRaf and BRAF and this may explain the observed cooperativity of cRaf and BRAF in cells responding to growth factor signals. Activating mutations in the BRAF gene are present in a large percentage of human malignant melanomas and in a proportion of colon cancers. The vast majority of these mutations result in a valine to glutamic acid change at residue 599 within the activation segment of BRAF.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAB1412331SAB1412337SAB1412336
assay

≥70% (SDS-PAGE)

assay

-

assay

-

assay

-

Gene Information

human ... BRAF(673)

Gene Information

human ... BRAF(673)

Gene Information

human ... BRAF(673)

Gene Information

human ... BRAF(673)

specific activity

63-85 nmol/min·mg

specific activity

-

specific activity

-

specific activity

-

recombinant

expressed in baculovirus infected Sf9 cells

recombinant

-

recombinant

-

recombinant

-

form

buffered aqueous glycerol solution

form

buffered aqueous solution

form

buffered aqueous solution

form

buffered aqueous solution

storage temp.

−70°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

9002-93-1

CAS No.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters