Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
25 mg | Check Cart for Availability | € 618,00 |
About This Item
Quality Segment
assay
≥98% (HPLC)
form
solid
color
off-white
solubility
DMSO: ~20 mg/mL, H2O: insoluble
storage temp.
2-8°C
SMILES string
Brc1ccc(NC(=O)c2cccc(c2)S(=O)(=O)Nc3ccc(Br)cc3)cc1
InChI
1S/C19H14Br2N2O3S/c20-14-4-8-16(9-5-14)22-19(24)13-2-1-3-18(12-13)27(25,26)23-17-10-6-15(21)7-11-17/h1-12,23H,(H,22,24)
InChI key
SJJKUNDBFWUAQB-UHFFFAOYSA-N
Biochem/physiol Actions
1 of 1
This Item | |||
|---|---|---|---|
| form solid | form solid | form solid | form solid |
| assay ≥98% (HPLC) | assay ≥98% (HPLC) | assay ≥98% (HPLC) | assay ≥98% (HPLC) |
| Quality Level 100 | Quality Level - | Quality Level 100 | Quality Level 100 |
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. −20°C | storage temp. 2-8°C |
| solubility DMSO: ~20 mg/mL, H2O: insoluble | solubility H2O: soluble ~10 mg/mL at 60 °C (Sonicate), DMSO: soluble 12 mg/mL | solubility DMSO: soluble >10 mg/mL, H2O: insoluble | solubility H2O: insoluble <2 mg/mL, DMSO: >5 mg/mL |
| color off-white | color light brown | color white to light tan | color brown |
signalword
Warning
hcodes
Hazard Classifications
Aquatic Chronic 4 - Eye Irrit. 2
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
![N-(4-Bromophenyl)-3-[[(4-bromophenyl)amino]sulfonyl]benzamide ≥98% (HPLC), solid](/deepweb/assets/sigmaaldrich/product/structures/120/254/f9829d51-31a0-469a-ab52-5e2f49b48d81/640/f9829d51-31a0-469a-ab52-5e2f49b48d81.png)
![cis-8-Hydroxy-3-(n-propyl)-1,2,3a,4,5,9b-hexahydro-1H-benz[e]indole hydrobromide ≥98% (HPLC), solid](/deepweb/assets/sigmaaldrich/product/structures/255/210/7ae0c9f3-835e-4c95-aa84-bb7695fe11ac/640/7ae0c9f3-835e-4c95-aa84-bb7695fe11ac.png)


