Skip to Content
Merck

Skip To

B2929

B-Lymphocyte Chemoattractant human

≥95% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture

Synonym(s):

B-Cell Attracting Chemokine 1, BCA-1, BLC

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 μg
Check Cart for Availability
€ 640,00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32
MDL number:
Biological source:
human
Recombinant:
expressed in E. coli
Assay:
≥95% (SDS-PAGE)
Form:
lyophilized powder
Mol wt:
apparent mol wt 8.7 kDa by SDS-PAGE (reducing)
Impurities:
endotoxin, tested

€ 640,00


Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human

Quality Segment

recombinant

expressed in E. coli

assay

≥95% (SDS-PAGE)

form

lyophilized powder

potency

0.1000 μg/mL

mol wt

apparent mol wt 8.7 kDa by SDS-PAGE (reducing)

packaging

pkg of 25 μg

storage condition

avoid repeated freeze/thaw cycles (Do not store in a frost-free freezer.)

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... CXCL13(10563)

Biochem/physiol Actions

BCA-1 is a potent chemoattractant for B lymphocytes, but not T lymphocytes, monocytes, or neutrophils.
BCA-1 is a potent chemoattractant for B lymphocytes, but not T lymphocytes, monocytes, or neutrophils. Its specific receptor BLR1 is a G protein-coupled receptor originally isolated from Burkitt′s lymphoma cells. Among cells of the hematopoietic lineages, the expression of BRL1, now designated CXCR5, is restricted to B lymphocytes and a subpopulation of T helper memory cells.
Potent chemoattractant for B lymphocytes, but not T lymphocytes, monocytes, or neutrophils.
The biological activity of human BLC/BCA-1 is measured by its ability to chemoattract mouse BaF/3 cells transfected with human CXCR5.

Physical form

Lyophilized from a 0.2 μm filtered solution in 35% acetonitrile and 0.1% TFA containing 50 μg bovine serum albumin per 1 μg of cytokine.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A0476B1434F135
recombinant

expressed in E. coli

recombinant

expressed in NSO cells

recombinant

expressed in CHO cells

recombinant

expressed in NSO cells

biological source

human

biological source

human

biological source

human

biological source

human

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

assay

≥95% (SDS-PAGE)

assay

>90% (SDS-PAGE)

assay

>95% (SDS-PAGE)

assay

≥97% (SDS-PAGE)

Quality Level

200

Quality Level

100

Quality Level

100

Quality Level

200

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters