Skip to Content
Merck
All Photos(2)

Key Documents

B1636

Sigma-Aldrich

Pentagastrin

≥95% (HPLC), powder, CCK2 receptor agonist

Synonym(s):

Peptavlon, Petogasrin, N-t-Boc-β-Ala-Trp-Met-Asp-Phe amide

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€207.00
100 MG
€687.00

€207.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
25 MG
€207.00
100 MG
€687.00

About This Item

Empirical Formula (Hill Notation):
C37H49N7O9S
CAS Number:
Molecular Weight:
767.89
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€207.00


Estimated to ship on29 May 2025


Request a Bulk Order

Product Name

Pentagastrin, ≥95% (HPLC), powder

Assay

≥95% (HPLC)

form

powder

storage condition

(Keep container tightly closed in a dry and well-ventilated place.)

color

white to off-white

solubility

DMF: 20 mg/mL, clear, colorless to faint yellow or tan

storage temp.

−20°C

SMILES string

CSCCC(NC(=O)C(Cc1c[nH]c2ccccc12)NC(=O)CCNC(=O)OC(C)(C)C)C(=O)NC(CC(O)=O)C(=O)NC(Cc3ccccc3)C(N)=O

InChI

1S/C37H49N7O9S/c1-37(2,3)53-36(52)39-16-14-30(45)41-28(19-23-21-40-25-13-9-8-12-24(23)25)34(50)42-26(15-17-54-4)33(49)44-29(20-31(46)47)35(51)43-27(32(38)48)18-22-10-6-5-7-11-22/h5-13,21,26-29,40H,14-20H2,1-4H3,(H2,38,48)(H,39,52)(H,41,45)(H,42,50)(H,43,51)(H,44,49)(H,46,47)

InChI key

NEYNJQRKHLUJRU-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Boc-Bal-Trp-Met-Asp-Phe-NH2

General description

Research area: Immunology & cytokines

Application

Pentagastrin has been used to bidirectionally modulate lateral nucleus of the amygdala (LA) activity.[1] It has also been used as a cholecystokinin-2 receptor (CCK2R) agonist to block the receptor binding for Ga-DOTA-MGS5 in AR42J rat pancreatic cell lines during cell uptake and receptor binding studies.[2]

Biochem/physiol Actions

Pentagastrin, a synthetic polypeptide is a gastrin analog. It contains the five C-terminal amino acids of gastrin.[3] It acts as a cholecystokinin-2 (CCK2) receptor/ cholecystokinin-B (CCKB) receptor agonist.[4] Pentagastrin stimulates the secretion of pepsin and gastric acid.[3] It also aids in the evaluation of gastric acid secretory function.[5] Pentagastrin exhibits panicogenic/ anxiogenic effects and thereby induces panic attacks in panic disorder (PD) patients.[4]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pentagastrin as a stimulant in routine clinical testing of gastric acid secretion.
S A Pedersen
Digestion, 3(2), 81-89 (1970-01-01)
Gilbert H Daniels
Thyroid : official journal of the American Thyroid Association, 21(11), 1199-1207 (2011-09-23)
Medullary thyroid carcinoma (MTC) is a rare thyroid malignancy with the potential for aggressive behavior. Measurement of serum calcitonin (Ct) in the thyroid nodule population is the most sensitive way to detect occult MTC. An important and controversial question is
Functional, Diagnostic and Therapeutic Aspects of Gastrointestinal Hormones
Ahmed M and Ahmed S
Gastroenterology Research and Practice, 233-244 (2019)
Anton A Hörmann et al.
Pharmaceuticals (Basel, Switzerland), 14(6) (2021-07-03)
The new minigastrin analog DOTA-MGS5 is a promising new candidate for targeting cholecystokinin-2 receptor (CCK2R)-expressing tumors. To enable the clinical translation of PET/CT imaging using 68Ga-labeled DOTA-MGS5, different quality and safety aspects need to be considered to comply with the
Géraldine Pina et al.
Clinical endocrinology, 78(3), 358-364 (2012-08-24)
To evaluate a second-generation assay for basal serum calcitonin (CT) measurements compared with the pentagastrin-stimulation test for the diagnosis of inherited medullary thyroid carcinoma (MTC) and the follow-up of patients with MTC after surgery. Recent American Thyroid Association recommendations suggest

Questions

1–4 of 4 Questions  
  1. How can Pentagastrin, Product B1636, solutions be stored?

    1 answer
    1. Pentagastrin is subject to oxidation in solution.  A pentagastrin solution in DMF can be stored frozen to protect the TRP and MET from oxidation. Further protection from oxidation can be insured if the solution is prepared with degassed solvent and the solution is stored under an inert gas such as argon or dry nitrogen gas.

      Helpful?

  2. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  3. What is the shelf-life of Pentagastrin, Product B1636?

    1 answer
    1. This product does not have any official shelf-life or expiration dating associated with it. The only date shown on the certificate of analysis will be the QC Release date. For customers whose systems require formal date management, use of a date one year from shipment is supported by our terms and conditions of supply.

      Helpful?

  4. What is Pentagastrin, Product B1636, soluble in?

    1 answer
    1. Pentagastrin is soluble in either dimethylformamide (DMF) or DMSO at a concentration of 20 mg/mL.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service