Skip to Content
Merck
All Photos(2)

Key Documents

S8820

Sigma-Aldrich

Protease Inhibitor Cocktail

SIGMA FAST, tablet, for the inhibition of serine, cysteine, aspartic and metalloproteases, For General Use

Synonym(s):

Protease Inhibitor Tablets, protease inhibitor

Sign Into View Organizational & Contract Pricing

Select a Size

2 TABS
€70.40
20 TABS
€637.00

€70.40


Estimated to ship on26 March 2025



Select a Size

Change View
2 TABS
€70.40
20 TABS
€637.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.77
form:
tablet
solubility:
water: 1 tablet/100mL, clear to hazy, colorless
storage temp.:
2-8°C

€70.40


Estimated to ship on26 March 2025


Product Name

SIGMAFAST Protease Inhibitor Tablets, For General Use

biological source

bovine lung
bovine milk

Quality Level

form

tablet

solubility

water: 1 tablet/100mL, clear to hazy, colorless

storage temp.

2-8°C

General description

SIGMAFAST Protease Inhibitor Tablets are a water-soluble tablet formulation containing a broad-spectrum cocktail of protease inhibitors, including AEBSF, E-64, bestatin, leupeptin, aprotinin, and EDTA. One tablet can make 100 mL of cocktail and can inhibit proteases present in up to 20 g of cell extract.
The use of SIGMAFAST Protease Inhibitor Tablets is not advised with HIS-Select® products or other immobilized metal affinity chromatography (IMAC) products. These tablets contain EDTA, which is a metal chelator and can potentially remove metal ions from these affinity products.

Specificity

Inhibits serine, cysteine, aspartic, and metalloproteases.

Application

SIGMAFAST Protease Inhibitor Tablets has been used:
  • to wash raw semen for membrane protein analysis[1]
  • in a solution to incubate CD4+ T cells to obtain total cell lysates for western blot analyses[2]
  • as a component of NP-40 buffer to homogenize the spinal cord tissue centered at the injury epicenter to conduct western blot analysis[3]
  • as a component in radio-immunoprecipitation assay (RIPA) buffer to homogenize the spinal cord tissue centered at the injury center to detect neuregulin-1 beta 1 (Nrg-1ß1) in spinal cord tissue samples with enzyme-linked immunosorbent assay (ELISA)[3]

These tablets have been optimized and tested for general use. They have been used in various applications, including collecting tracheae in combination with ice-cold radioimmunoprecipitation assay buffer to determine the tracheal basal cell fate, and homogenizing rat lung tissues for western blot analysis.

Features and Benefits

  • Broad-spectrum protease inhibition: inhibits serine, cysteine, aspartic, and metalloproteases, making it a versatile solution for protease inhibition in various applications.
  • Water-soluble tablet formulation: easy to use and prepare, with one tablet making 100 mL of cocktail.
  • Contains a cocktail of protease inhibitors: provides a comprehensive and effective solution for protease inhibition.
  • Tested and optimized for general use: proven performance and reliability for a wide range of applications.

Components

AEBSF, 2 mM
Aprotinin, 0.3 μM
Bestatin, 130 μM
EDTA, 1 mM
E-64, 14 μM
Leupeptin, 1 μM

Quantity

One tablet makes 100 mL of cocktail. One mL is recommended for the inhibition of proteases equivalent to 1 mg of USP pancreatin. One tablet is recommended for the inhibition of proteases present in a maximum of 20 g of cell extract.

Physical form

Tablet that is water soluble.

Other Notes

For R&D use only. Not for drug, household, or other uses. Please consult the Safety Data Sheet for information regarding hazards and safe handling practices.

Legal Information

HIS-Select is a registered trademark of Merck KGaA, Darmstadt, Germany
SIGMAFAST is a trademark of Sigma-Aldrich Co. LLC

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Immunometabolic profiling of T cells from patients with relapsing-remitting multiple sclerosis reveals an impairment in glycolysis and mitochondrial respiration
La Rocca C, et al.
Journal of biological and chemical chronicles, 77, 39-46 (2017)
Tran Hong Nha Nguyen et al.
Molecular & cellular proteomics : MCP, 11(11), 1140-1155 (2012-07-31)
Root hairs are single hair-forming cells on roots that function to increase root surface area, enhancing water and nutrient uptake. In leguminous plants, root hairs also play a critical role as the site of infection by symbiotic nitrogen fixing rhizobia
Nihar J Mehta et al.
Free radical biology & medicine, 101, 44-52 (2016-11-05)
The dopamine oxidation product cysteinyl-dopamine has attracted attention as a contributor to the death of dopaminergic neurons in Parkinson's disease. Treatment of cysteinyl-dopamine with hypochlorite yields an even more cytotoxic product. This product has potent redox-cycling activity and initiates production
Artavazd Tadevosyan et al.
The Journal of biological chemistry, 285(29), 22338-22349 (2010-05-14)
Angiotensin-II (Ang-II) from extracardiac sources and intracardiac synthesis regulates cardiac homeostasis, with mitogenic and growth-promoting effects largely due to altered gene expression. Here, we assessed the possibility that angiotensin-1 (AT1R) or angiotensin-2 (AT2R) receptors on the nuclear envelope mediate effects
Protein content of equine seminal plasma and sperm plasma membrane in subfertile stallions
Guasti PN, et al.
Journal of equine veterinary science, 34(1), 84-85 (2014)

Articles

While aprotinin and bovine pancreatic trypsin inhibitor (BPTI) are the same protein sequence, the term aprotinin is typically used when describing the protein derived from bovine lung.

Related Content

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Questions

1–8 of 8 Questions  
  1. How long does the cocktail keep once a tablet is reconstituted in 10mL of water at 2-8°C?

    1 answer
    1. The reconstituted protease inhibitor solution is stable for at least 2 weeks at 2-8 °C. Please see the datasheet below for more information:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/177/562/s8820bul-ms.pdf

      Helpful?

  2. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  3. What is the concentration of the inhibitors in Product S8820, SIGMAFAST™ Protease Inhibitor Tablets?

    1 answer
    1. When reconstituted (one tablet in 100 ml water), concentrations of the component inhibitors are as follows:AEBSF      2 mME-64          14 μMBestatin     130 μMLeupeptin   0.9 μMAprotinin    0.3 μMEDTA        1 mM

      Helpful?

  4. How much of Product S8820, SIGMAFAST™ Protease Inhibitor Tablets, needs to be used in cell lysates?

    1 answer
    1. One tablet is recommended for the inhibition of proteases present in a maximum of 20 grams of cell extract.

      Helpful?

  5. Can Product S8820, SIGMAFAST™ Protease Inhibitor Tablets, be used with with HIS-Select™ products or any other immobilized metal affinity chromatography (IMAC) products?

    1 answer
    1. The tablets contain EDTA, a metal chelator, which may remove metal ions from these affinity products. The suggested protease inhibitor for the HIS-Select products is either Product No. S8830 (water soluble tablet) or P8849 (DMSO solution).

      Helpful?

  6. Can Product S8820, SIGMAFAST™ Protease Inhibitor Tablets, be used with ELISA?

    1 answer
    1. Yes, because it is water based.

      Helpful?

  7. How should I dissolve Product S8820, SIGMAFAST™ Protease Inhibitor Tablets?

    1 answer
    1. You can dissolve the product in 10 ml of water to make a 10X stock solution and then dilute 1:10 for use.

      Helpful?

  8. What is the difference between Protease Inhibitors S8820 and P2714?

    1 answer
    1. S8820 is the tablet version of P2714.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service