Skip to Content
Merck

Skip To

B1379

5-Bromo-4-chloro-3-indolyl sulfate potassium salt

sulfatase substrate, ≥98% (TLC), powder

Synonym(s):

5-bromo-4-chloroindoxyl sulfate potassium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 mg
Check Cart for Availability
€ 236,00
250 mg
Check Cart for Availability
€ 316,00
€ 268,60

About This Item

Empirical Formula (Hill Notation):
C8H4BrClKNO4S
CAS Number:
Molecular Weight:
364.64
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83
MDL number:

€ 236,00


Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

5-Bromo-4-chloro-3-indolyl sulfate potassium salt, sulfatase substrate

Quality Segment

assay

≥98% (TLC)

form

powder

solubility

water: 50 mg/mL, clear to slightly hazy

storage temp.

−20°C

SMILES string

[K+].[O-]S(=O)(=O)Oc1c[nH]c2ccc(Br)c(Cl)c12

InChI

1S/C8H5BrClNO4S.K/c9-4-1-2-5-7(8(4)10)6(3-11-5)15-16(12,13)14;/h1-3,11H,(H,12,13,14);/q;+1/p-1

InChI key

BONBCMDYPZGTEU-UHFFFAOYSA-M

General description

A histochemical substrate for aryl-sulfatase.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
N3877P4154H8759
assay

≥98% (TLC)

assay

≥98% (TLC)

assay

≥98% (TLC)

assay

≥98% (HPLC)

solubility

water: 50 mg/mL, clear to slightly hazy

solubility

water: 50 mg/mL, clear, colorless to very faintly greenish-yellow (to Very Light Yellow)

solubility

water: 4.80-5.20 mg/mL, clear to hazy, colorless to faintly yellow

solubility

H2O: 50 mg/mL

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

form

powder

form

powder

form

lyophilized powder

form

powder

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

2-8°C


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters