Skip to Content
Merck

Skip To

B1185

Bioreactor pH Adjustment Solution

liquid, sterile-filtered, suitable for cell culture

Synonym(s):

Bioreactor buffer solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 L
Check Cart for Availability
€ 226,00

About This Item

NACRES:
NA.75
UNSPSC Code:
12352107

€ 226,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

non-animal source

Quality Segment

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

shipped in

ambient

storage temp.

room temp

Application

Concentrated carbonate/bicarbonate solution designed specifically to adjust pH in bioreactor applications.

Features and Benefits

Using this buffered solution to adjust the pH in bioreactors minimizes detrimental localized pH effects and the osmolality impact compared to using NaOH.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
H6909A9576A8412
form

liquid

form

liquid

form

liquid

form

liquid

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

biological source

non-animal source

biological source

non-animal source

biological source

bovine

biological source

bovine

sterility

sterile-filtered

sterility

sterile-filtered

sterility

sterile-filtered

sterility

sterile-filtered

storage temp.

room temp

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

2-8°C

shipped in

ambient

shipped in

dry ice

shipped in

ambient

shipped in

ambient


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters