Skip to Content
Merck

Skip To

AV46751

Anti-VAMP5 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Vesicle-associated membrane protein 5 (Myobrevin)

Sign In to View Organizational & Contract Pricing.

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
2


biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

13 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... VAMP5(10791)

Immunogen

Synthetic peptide directed towards the middle region of human VAMP5

Application

Anti-VAMP5 antibody produced in rabbit is suitable for western blotting at a concentration of 5μg/mL.

Biochem/physiol Actions

VAMP5 (vesicle-associated membrane protein 5) gene encodes a 116 amino acid containing single-pass type IV membrane protein that belongs to the synaptobrevin family and the SNARE superfamily. It plays a pivotal role vesicle trafficking events that are associated with myogenesis like- myoblast fusion and/or GLUT4 trafficking. VAMP5 protein is localized in skeletal muscle, heart, spleen, lung, liver, and kidney tissue and facilitates the membrane trafficking in skeletal and cardiac muscle.

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Other Notes

Synthetic peptide located within the following region: IRYRICVGLVVVGVLLIILIVLLVVFLPQSSDSSSAPRTQDAGIASGPGN

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
HPA035082SAB2107401SAB5701461
antibody form

IgG fraction of antiserum

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

-

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

UniProt accession no.

O95183

UniProt accession no.

O95183

UniProt accession no.

P23763

UniProt accession no.

P23763

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Questions

Reviews

No rating value

Active Filters