Skip to Content
Merck

Skip To

AV42101

Anti-SQLE antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-FLJ30795, Anti-Squalene epoxidase

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μL
Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)
€ 507,00

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, WB
Citations:
-

€ 507,00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

39 kDa

species reactivity

human, bovine, rabbit, mouse, sheep

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SQLE(6713)

General description

Squalene epoxidase (SQLE) is an NADPH-dependent flavoprotein monooxygenase that oxidizes squalene to 2,3,-oxidosqualene (squalene epoxide) which is the first oxygenation and rate-limiting step in sterol, ergosterol and cholesterol, biosynthesis.

Immunogen

Synthetic peptide directed towards the C terminal region of human SQLE

Application

Anti-SQLE polyclonal antibody is used to tag squalene epoxidase for detection and quantitation by Western blotting and in plasma by immunohistochemical (IHC) techniques. It is used as a probe to determine the roles of squalene epoxidase in sterol, ergosterol and cholesterol, biosynthesis.

Biochem/physiol Actions

Anti-SQLE polyclonal antibody reacts with human, mouse, rat, pig, zebrafish, bovine, and canine squalene epoxidases.
Squalene epoxidase catalyzes the first oxygenation step in sterol biosynthesis and is thought to be one of the rate-limiting enzymes in this pathway.

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Other Notes

Synthetic peptide located within the following region: KKSFYWARKTSHSFVVNILAQALYELFSATDDSLHQLRKACFLYFKLGGE

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
HPA018038SAB1406474HPA045520
Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

biological source

rabbit

biological source

rabbit

biological source

mouse

biological source

rabbit

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

purified immunoglobulin

antibody form

affinity isolated antibody

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

species reactivity

human, bovine, rabbit, mouse, sheep

species reactivity

human

species reactivity

human

species reactivity

human

Gene Information

human ... SQLE(6713)

Gene Information

human ... SQLE(6713)

Gene Information

human ... SQLE(6713)

Gene Information

human ... SRL(6345)


Still not finding the right product?

or

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters