Select a Size
| Pack Size | SKU | Availability | Price |
|---|
About This Item
biological source
rabbit
Quality Segment
antibody form
Ig fraction of antiserum
clone
monoclonal
description
For In Vitro Diagnostic Use in Select Regions (See Chart)
form
buffered aqueous solution
technique(s)
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:200
shipped in
wet ice
storage temp.
2-8°C
visualization
cytoplasmic
General description
Physical form
Analysis Note
![]() IVD | ![]() IVD | ![]() IVD | ![]() RUO |
Other Notes
For Technical Service please contact: 800-665-7284 or email: [email protected]
1 of 1
This Item | |||
|---|---|---|---|
| antibody form Ig fraction of antiserum | antibody form Ig fraction of antiserum | antibody form affinity isolated antibody | antibody form Ig fraction of antiserum |
| clone monoclonal | clone monoclonal | clone polyclonal | clone EP109, monoclonal |
| biological source rabbit | biological source rabbit | biological source rabbit | biological source rabbit |
| shipped in wet ice | shipped in wet ice | shipped in wet ice | shipped in wet ice |
| form buffered aqueous solution | form buffered aqueous solution | form buffered aqueous glycerol solution | form buffered aqueous solution |
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. −20°C | storage temp. 2-8°C |
Still not finding the right product?
Explore all of our products under Prostein (P501S, SLC45A3) (EP381) Rabbit Monoclonal Primary Antibody
Storage Class
10 - Combustible liquids
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| AC-0335 | 04061838508652 |





