Skip to Content
Merck

Skip To

A9847

Angiopoietin-2 human

>97% (SDS-PAGE), recombinant, expressed in NSO cells, lyophilized powder

Synonym(s):

Ang-2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 μg
Check Cart for Availability
€ 1.000,00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32
MDL number:
Biological source:
human
Recombinant:
expressed in NSO cells
Assay:
>97% (SDS-PAGE)
Form:
lyophilized powder
Mol wt:
66 kDa by SDS-PAGE (reducing and non-reducing conditions), 66 kDa by SDS-PAGE
Impurities:
endotoxin, tested

€ 1.000,00


Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human

Quality Segment

recombinant

expressed in NSO cells

assay

>97% (SDS-PAGE)

form

lyophilized powder

mol wt

66 kDa by SDS-PAGE (reducing and non-reducing conditions), 66 kDa by SDS-PAGE

packaging

pkg of 25 μg

storage condition

avoid repeated freeze/thaw cycles (Do not store in a frost-free freezer.)

impurities

endotoxin, tested

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... ANGPT2(285)

Biochem/physiol Actions

This ligand is a naturally occurring inhibitor of angiopoietin-1 and Tie-2 both in vivo and in vitro, and has important roles in embryogenic angiogenesis and maintaining the integrity of adult vasculature.

Physical form

Lyophilized from a 0.2 μm filtered solution in 20 mM MOPS, 500 mM NaCl, 0.1% PEG-8000, 0.2% CHAPS, pH 7.0 containing 50 μg bovine serum albumin per 1 μg of cytokine.

Analysis Note

The biological activity is measured a competitive binding assay.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
E0278A0476B1434
recombinant

expressed in NSO cells

recombinant

expressed in NSO cells

recombinant

expressed in NSO cells

recombinant

expressed in CHO cells

biological source

human

biological source

human

biological source

human

biological source

human

assay

>97% (SDS-PAGE)

assay

>90% (SDS-PAGE)

assay

>90% (SDS-PAGE)

assay

>95% (SDS-PAGE)

Quality Level

200

Quality Level

200

Quality Level

100

Quality Level

100

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

storage condition

avoid repeated freeze/thaw cycles (Do not store in a frost-free freezer.)

storage condition

avoid repeated freeze/thaw cycles

storage condition

avoid repeated freeze/thaw cycles (Do not store in a frost-free freezer.)

storage condition

avoid repeated freeze/thaw cycles (Do not store in a frost-free freezer.)


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters