Sign In to View Organizational & Contract Pricing.
About This Item
Empirical Formula (Hill Notation):
C16H14N4O4S
CAS Number:
Molecular Weight:
358.37
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352202
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
assay
≥98% (HPLC)
form
powder
color
yellow to brown
solubility
DMSO: ≥30 mg/mL
storage temp.
2-8°C
SMILES string
COc1ccc2[nH]c(SCC(=O)Nc3ccc(cc3)[N+]([O-])=O)nc2c1
InChI
1S/C16H14N4O4S/c1-24-12-6-7-13-14(8-12)19-16(18-13)25-9-15(21)17-10-2-4-11(5-3-10)20(22)23/h2-8H,9H2,1H3,(H,17,21)(H,18,19)
InChI key
LYQDSNOFTIZWAX-UHFFFAOYSA-N
Biochem/physiol Actions
AG-09/1 is a selective formyl peptide receptor 1 (FPR1) agonist; it activates chemotaxis in human neutrophils.
AG-09/1 is a selective formyl peptide receptor 1 (FPR1) agonist; it activates chemotaxis in human neutrophils. Formyl peptide recetors (FPRs) are GPCRs mainly expressed in phagocytic leukocytes but with lower expression in many other cell types. Formyl peptides, act as Alarmins and are released from bacteria and damaged mitochondria, serving as chemoattractants for phagocyte recruitment to sites of inflammation, resulting in immune response. Because the endogenous ligands for FPRs are peptides and arachidonic acid metabolites, there are few small molecule tools for these receptors. A recent HTS of commercially available libraries yielded several FPR-1 selective agonists, of which AG-091 was the most potent. AG-091 induced intracellular calcium flux in FPR-1- but not FPR-2-expressing cells. It also caused chemotaxis and intracellular calcium flux in human leukocytes. AG-091 is a valuable tool for studying FPR1 function in immune and other disease states.
signalword
Danger
hcodes
Hazard Classifications
Eye Dam. 1
Storage Class
11 - Combustible Solids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
