Sign In to View Organizational & Contract Pricing.
About This Item
Linear Formula:
NH2COCH2CH(NH2)COOH · H2O
CAS Number:
Molecular Weight:
150.13
EC Number:
221-521-1
UNSPSC Code:
12352200
PubChem Substance ID:
Beilstein/REAXYS Number:
1723525
MDL number:
assay
≥98% (TLC)
mp
220 °C (dec.) (lit.)
SMILES string
O.NC(CC(N)=O)C(O)=O
InChI
1S/C4H8N2O3.H2O/c5-2(4(8)9)1-3(6)7;/h2H,1,5H2,(H2,6,7)(H,8,9);1H2
InChI key
RBMGJIZCEWRQES-UHFFFAOYSA-N
Storage Class
13 - Non Combustible Solids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
