Skip to Content
Merck

Skip To

A7477

Anti-ADAM-33, Catalytic Domain antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-A Disintegrin And Metalloproteinase-33

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12352203
MDL number:


biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human

concentration

~1 mg/mL

technique(s)

western blot: 1:1,000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... ADAM33(80332)

General description

A Disintegrin And Metalloproteinase-33 (ADAM-3) is a widely expressed protein that regulates cell-attachment by binding to integrins via cysteine-rich domains[1]. ADAM-33 has also been associated with asthma susceptibility[2]. Rabbit Anti-ADAM-33, Catalytic Domain antibody binds to the amino end of active human ADAM-33.
By immunoblotting against the reduced protein, the antibody identifies bands at 99 kDa (minor band), 58 kDa (major band), and cleaved products in cell lysates.

Immunogen

synthetic peptide corresponding to the amino end of active human ADAM-33.

Application

Rabbit Anti-ADAM-33, Catalytic Domain antibody can be used for westen blot applications at 1:1,000.

Physical form

Solution in phosphate buffered saline containing 50% glycerol and 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content


Tsuyoshi Yoshinaka et al.
Gene, 282(1-2), 227-236 (2002-01-30)
The ADAM family of membrane-anchored proteins has a unique domain structure, with each containing a disintegrin and metalloprotease (ADAM) domain. We have isolated mouse and human cDNAs encoding a novel member of the ADAM family. The mouse and human predicted
Paul Van Eerdewegh et al.
Nature, 418(6896), 426-430 (2002-07-12)
Asthma is a common respiratory disorder characterized by recurrent episodes of coughing, wheezing and breathlessness. Although environmental factors such as allergen exposure are risk factors in the development of asthma, both twin and family studies point to a strong genetic