Skip to Content
Merck

Skip To

A6855

Arachidonoyl-AMC

fatty acid amide hydrolase substrate, 99% (HPLC), solid

Synonym(s):

AAMCA

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 mg
Check Cart for Availability
€ 209,00
€ 177,65
25 mg
Check Cart for Availability
€ 748,00

About This Item

Empirical Formula (Hill Notation):
C30H39NO3
Molecular Weight:
461.64
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32
MDL number:

€ 177,65

List Price€ 209,00Save 15%
Web-Only Promotion

Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Arachidonoyl-AMC, fatty acid amide hydrolase substrate

Quality Segment

assay

99% (HPLC)

form

solid

storage condition

under inert gas

solubility

DMSO: soluble

storage temp.

−20°C

SMILES string

CCCCC\C=C/C\C=C/C\C=C/C\C=C/CCCC(=O)Nc1ccc2C(C)=CC(=O)Oc2c1

InChI

1S/C30H39NO3/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-29(32)31-26-21-22-27-25(2)23-30(33)34-28(27)24-26/h7-8,10-11,13-14,16-17,21-24H,3-6,9,12,15,18-20H2,1-2H3,(H,31,32)/b8-7-,11-10-,14-13-,17-16-

InChI key

IQPBUDMTXHWSHB-ZKWNWVNESA-N

Application

Fluorogenic substrate used in a selective and sensitive assay of fatty acid amide hydrolase (FAAH).

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
B7431E1904S8197
assay

99% (HPLC)

assay

≥99%

assay

>99% (HPLC)

assay

>99% (HPLC)

solubility

DMSO: soluble

solubility

DMSO: soluble, ethanol: soluble

solubility

DMSO: soluble 22 mg/mL, H2O: soluble 8 mg/mL

solubility

DMSO: >20 mg/mL, H2O: insoluble

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

form

solid

form

solid

form

solid

form

solid

storage temp.

−20°C

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

2-8°C

storage condition

under inert gas

storage condition

-

storage condition

-

storage condition

-


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters