Skip to Content
Merck
All Photos(3)

Key Documents

A6814

Sigma-Aldrich

α-Amylase from Bacillus sp.

powder, ≥400 units/mg protein (Lowry)

Synonym(s):

1,4-α-D-Glucan-glucanohydrolase

Sign Into View Organizational & Contract Pricing

Select a Size

1000000 UNITS
€188.00
5000000 UNITS
€703.00
25000000 UNITS
€2,560.00

€188.00


Estimated to ship on13 April 2025



Select a Size

Change View
1000000 UNITS
€188.00
5000000 UNITS
€703.00
25000000 UNITS
€2,560.00

About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
eCl@ss:
32160410
NACRES:
NA.54

€188.00


Estimated to ship on13 April 2025


biological source

Bacillus sp.

Quality Level

form

powder

specific activity

≥400 units/mg protein (Lowry)

mol wt

58-62 kDa

solubility

H2O: soluble 1.0 mg/mL

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

α-Amylase is used to hydrolyze α bonds of α-linked polysaccharides, such as starch and glycogen. α-Amylase, from Sigma, has been used in various plant studies, such as metabolism studies in Arabidopsis [1]. α-has also been also used to study the ability of corn and starch to protect enzyme activity.[2] The enzyme has been used to compare the effects of bacterial and fungal α-amylase and scalded flour on bread quality and shelflife. It has also been used to determine the optimal levels of these components in order to achieve maximum volume and bread freshness.[3] Furthermore, it has been used to digest hydroxypropylated starch in Korean waxy rice cake to retard retrogradation.[4]

Biochem/physiol Actions

α-Amylase hydrolyzes the α-(1,4) glucan linkages in polysaccharides of three or more α-(1,4) linked D-glucose units. Natural substrates such as starch and glycogen are broken down into glucose and maltose. α -amylase, isolated from Bacillus subtilis is a dimer and has a monomer molecular weight of 48.9 kDa. α-Amylase is a metalloprotein and contains atleast one mol of calcium ion. The presence of calcium ion increases the stability of the enzyme towards denaturation by heat, acid, or urea. The pH range for activity of this product is 5.0 to 7.5, with the optimum pH range being 6.0-7.0. This product is stable from pH 5.0 to 10.0. The optimum temperature range is 65-75 °C. The effective temperature range is up to 90 °C.[5]

Quality

Contains starch as an extender.

Unit Definition

One unit will liberate 1.0 mg of maltose from starch in 3 min at pH 6.9 at 20 °C.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Utilization of Hydroxypropylated Waxy Rice and Corn Starches in Korean Waxy Rice Cake to Retard Retrogradation
Han J-A, et al.
Cereal Chem., 82(1), 88-92 (2005)
Comparison of the effects of microbial α-amylases and scalded flour on bread quality
Hopek M, et al.
Acta Scientiarum Polonorum. Technologia Alimentaria, 5(1), 97-106 (2006)
Theoretical and Experimental Studies of the Effects of Heat, EDTA, and Enzyme Concentration on the Inactivation Rate of α-Amylase from Bacillus sp.
Lecker DN and Khan A
Biotechnol. Progress, 12(5), 713-717 (1996)
Usage of Enzyme Substrate to Protect the Activities of Cellulase, Protease and α-Amylase in Simulations of Monogastric Animal and Avian Sequential Total Tract Digestion
Wang HT and Hsu JT
Asian-Australasian Journal of Animal Sciences, 19(8), 1164-1173 (2006)
Sinéad Lordan et al.
Food chemistry, 141(3), 2170-2176 (2013-07-23)
To date, numerous studies have reported on the antidiabetic properties of various plant extracts through inhibition of carbohydrate-hydrolysing enzymes. The objective of this research was to evaluate extracts of seaweeds for α-amylase and α-glucosidase inhibitory effects. Cold water and ethanol

Protocols

Follow our procedure for the determination of alpha-Amylase activity. This enzymatic assay of a-Amylase guides you through the entire process and necessary calculations.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service