Skip to Content
Merck
All Photos(1)

Documents

N1519

Sigma-Aldrich

4-Nitrophenyl α-D-maltopentaoside

≥98%

Synonym(s):

4-Nitrophenyl α-D-penta-(1→4)-glucopyranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H55NO28
CAS Number:
Molecular Weight:
949.81
Beilstein:
4839501
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:

Assay

≥98%

form

powder

solubility

water: 50 mg/mL, clear, colorless to light yellow

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@H](O[C@H]2[C@H](O)[C@@H](O)[C@H](O[C@@H]2CO)O[C@H]3[C@H](O)[C@@H](O)[C@H](O[C@@H]3CO)O[C@H]4[C@H](O)[C@@H](O)[C@H](O[C@@H]4CO)O[C@H]5[C@H](O)[C@@H](O)[C@H](O[C@@H]5CO)Oc6ccc(cc6)[N+]([O-])=O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C36H55NO28/c38-5-12-17(43)18(44)23(49)33(57-12)62-29-14(7-40)59-35(25(51)20(29)46)64-31-16(9-42)61-36(27(53)22(31)48)65-30-15(8-41)60-34(26(52)21(30)47)63-28-13(6-39)58-32(24(50)19(28)45)56-11-3-1-10(2-4-11)37(54)55/h1-4,12-36,38-53H,5-9H2/t12-,13-,14-,15-,16-,17-,18+,19-,20-,21-,22-,23-,24-,25-,26-,27-,28-,29-,30-,31-,32+,33-,34-,35-,36-/m1/s1

InChI key

YXGBAQKCCMQLGH-MYPSSPKESA-N

Looking for similar products? Visit Product Comparison Guide

Substrates

Chromogenic substrate for α-amylase.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C S Elbin et al.
Clinical chemistry, 39(1), 112-118 (1993-01-01)
We describe a reagent for measuring alpha-amylase (EC 3.2.1.1) activity in serum with use of a thexyldimethylsilyl ether of p-nitrophenyl-alpha-D-maltoheptaoside (SB7) as substrate. This substrate differs from Genzyme's benzylidene-blocked p-nitrophenylmaltoheptaoside substrate (B-PNPG7). The reagent, optimized for the characteristics of the
T Usui et al.
Analytical biochemistry, 202(1), 61-67 (1992-04-01)
Enzymatic modification at the nonreducing end D-glucosyl residue of p-nitrophenyl alpha-maltopentaoside was developed by using the transglycosylation of beta-D-galactosidase from Bacillus circulans. The enzyme regioselectively synthesized p-nitrophenyl 4(5)-O-beta-D-galactosyl-alpha-maltopentaoside (a yield of 12.0% based on the amount of p-nitrophenyl alpha-maltopentaoside added)
J Sumitani et al.
The Biochemical journal, 350 Pt 2, 477-484 (2000-08-19)
The alpha-amylase from Bacillus sp. no. 195 (BAA) consists of two domains: one is the catalytic domain similar to alpha-amylases from animals and Streptomyces in the N-terminal region; the other is the functionally unknown domain composed of an approx. 90-residue
E H Ajandouz et al.
Carbohydrate research, 268(2), 267-277 (1995-03-17)
The catalytic efficiency (kcat/Km) and the cleaved bond distribution for the nitrophenylated maltooligosaccharides, p-NPGlcn (2 < or = n < or = 7) hydrolysed by porcine pancreatic alpha-amylase isozymes I and II were determined. The subsite affinities (Ai) were calculated
Sangeun Park et al.
Bioorganic & medicinal chemistry letters, 21(8), 2441-2444 (2011-03-15)
We constructed a library of sugar-dipeptide conjugate to find out the best complementary against hydrophobic pocket of α-glucosidase. The best substrate showed 150-fold improved K(m) value relative p-acetaminophenyl-α-D-glucopyranoside for α-glucosidase from Bacillus stearothermophillus. Using information from the complementary, we synthesized

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service