Skip to Content
Merck

Skip To

A6553

DL-2-Amino-5-phosphonovaleric acid lithium salt

~95%

Synonym(s):

DL-2-Amino-5-phosphonopentanoic acid, AP-5

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
C5H11NO5PLi
CAS Number:
Molecular Weight:
203.06
NACRES:
NA.26
PubChem Substance ID:
UNSPSC Code:
12352106
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

~95%

Quality Level

form

powder

technique(s)

ligand binding assay: suitable

color

white to off-white

SMILES string

[Li].NC(CCCP(O)(O)=O)C(O)=O

InChI

1S/C5H12NO5P.Li.H/c6-4(5(7)8)2-1-3-12(9,10)11;;/h4H,1-3,6H2,(H,7,8)(H2,9,10,11);;

InChI key

QIVHTKSFMZBERS-UHFFFAOYSA-N

Gene Information

Biochem/physiol Actions

Anticonvulsant; potent and selective antagonist for NMDA receptors.
DL-2-Amino-5-phosphonovaleric acid is a selective N-methyl-d-aspartate (NMDA) receptor antagonist used frequently in studies of neurological processes

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A8054A3785A7502
form

powder

form

powder

form

powder

form

powder

technique(s)

ligand binding assay: suitable

technique(s)

ligand binding assay: suitable

technique(s)

ligand binding assay: suitable

technique(s)

ligand binding assay: suitable

assay

~95%

assay

≥98% (TLC)

assay

≥98%

assay

≥99% (HPLC)

Quality Level

100

Quality Level

100

Quality Level

300

Quality Level

200

color

white to off-white

color

white

color

-

color

white to off-white

Gene Information

rat ... Grik1(29559), Grin2a(24409), Grin2b(24410)

Gene Information

mouse ... Grin2a(14811)
rat ... Grik1(29559), Grin2a(24409), Grin2b(24410), Grin2c(24411), Grin2d(24412)

Gene Information

-

Gene Information

-


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Jessica M André et al.
Neuropharmacology, 60(4), 617-625 (2010-12-21)
NMDA glutamate receptors (NMDARs) and nicotinic acetylcholine receptors (nAChRs) are both involved in learning and synaptic plasticity. Increasing evidence suggests processes mediated by these receptors may interact to modulate learning; however, little is known about the neural substrates involved in
A Osorio-Espinoza et al.
Neuroscience, 176, 20-31 (2011-01-05)
The globus pallidus, a neuronal nucleus involved in the control of motor behavior, expresses high levels of histamine H(3) receptors (H(3)Rs) most likely located on the synaptic afferents to the nucleus. In this work we studied the effect of the
Thomas K Fung et al.
Synapse (New York, N.Y.), 65(7), 677-686 (2010-12-31)
Synaptic plasticity may depend not only on the afferent fibers but also on the recipient structure. The medial perforant path (MPP) from the entorhinalcortex projects to both the dentate gyrus (DG) and CA3, resulting in excitatory postsynaptic potentials (EPSPs) in



Global Trade Item Number

SKUGTIN
A6553-25MG04061833382462
A6553-10MG04061832415024

Questions

Reviews

No rating value

Active Filters